Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-Carbobenzyloxy-4-amino-2-hydroxybutyric acid, 96%, Thermo Scientific™

Catalog No. AC342440250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC342440250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC342440250 Supplier Thermo Scientific Chemicals Supplier No. 342440250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 40371-50-4
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C12H14NO5
MDL Number MFCD00209957
Synonym s-n-carbobenzyloxy-4-amino-2-hydroxybutyric acid, s-n-cbz-4-amino-2-hydroxybutyric acid, cbz-haba, 2s-4-benzyloxy carbonyl amino-2-hydroxybutanoic acid, s-+-z-4-amino-2-hydroxybutyric acid, s,4-benzyloxycarbonylamino-2-hyroxybutyric acid, 2s-2-hydroxy-4-phenylmethoxy carbonylamino butanoic acid, pubchem11934, s-2-hydroxy-4-phenylmethoxy carbonyl amino butyric acid, acidol-4benzyloxycarbonyl-amino-2hydroxy butyric
InChI Key ULKOBRDRCYROKY-JTQLQIEISA-M
SMILES O[C@@H](CCNC(=O)OCC1=CC=CC=C1)C([O-])=O
IUPAC Name (2S)-4-{[(benzyloxy)carbonyl]amino}-2-hydroxybutanoate
Molecular Weight (g/mol) 252.25
PubChem CID 2733930
Formula Weight 253.25
Percent Purity ≥95%
Chemical Name or Material (S)-N-Carbobenzyloxy-4-amino-2-hydroxybutyric acid, 96%
Show More Show Less
EINECSNumber 254-892-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.