Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (S)-N-FMOC-Octylglycine, 95%

Catalog No. AC361490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC361490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC361490010 Supplier Thermo Scientific Chemicals Supplier No. 361490010
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 193885-59-5 | C25H31NO4 | 409.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 193885-59-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 142.6°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C25H31NO4
Quantity 1 g
MDL Number MFCD01311753
Synonym fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
InChI Key LSMLSLHVRMSYPT-QHCPKHFHSA-N
SMILES CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
Molecular Weight (g/mol) 409.52
PubChem CID 23257324
Formula Weight 409.52
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-Octylglycine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.