Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-N-Fmoc-Piperidine-2-Carboxylic Acid

Catalog No. 7064345G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-643-45G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-643-45G Supplier Sigma Aldrich Supplier No. 7064345G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-N-Fmoc-Piperidine-2-Carboxylic Acid

Specifications

CAS 12170-97-7
Melting Point 190°C
Molecular Formula C12H18Cl2Ru
Linear Formula C12H18Cl2Ru
MDL Number MFCD08459354
Quantity 500 mg
Synonym Dichloro(dodeca-2,6,10-triene-1,12-diyl)ruthenium(IV); Dichloro-2,6,10-dodecatrienylene-ruthenium; Dichloro[(1,2,3,6,7,10,11,12-eta)-2,6,10-dodecatriene-1,12-diyl]-ruthenium
Molecular Weight (g/mol) 334.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.