Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-N,N-Dimethyl-1-Ferrocenylethylamine

Catalog No. 402411ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-241-1ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-241-1ML Supplier Sigma Aldrich Supplier No. 402411ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine can be used:As a starting material in the synthesis of chiral diferrocenylphosphine-diamines applicable in catalysis.As a model chiral probe in the study of enantiodiscrimination ability of chiral ionic liquids using cyclic voltammetry.

Specifications

CAS 15278-97-4
Density 0.938 g/mL at 25°C
Linear Formula (CH3)3PAuCl
MDL Number MFCD00080961
Quantity 25 mL
Synonym (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.