Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine 97.0+%, TCI America™
SDP

Catalog No. D2529200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2529200MG 200 mg
D25291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2529200MG Supplier TCI America Supplier No. D2529200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31886-57-4
Molecular Formula C14H19FeN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00066272
Synonym (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene
IUPAC Name (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
SMILES [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1

Specifications

Boiling Point 121°C
Quantity 200 mg
Formula Weight 257.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.