Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N,N-Dimethyl-1-[(R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine 95.0+%, TCI America™
SDP

Catalog No. D2538100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2538100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2538100MG Supplier TCI America Supplier No. D2538100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55650-59-4
Molecular Formula C38H37FeNP2
MDL Number MFCD00075283
Synonym (S)-(R)-BPPFA

Specifications

Melting Point 142°C
Quantity 100 mg
Formula Weight 625.51
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N,N-Dimethyl-1-[(R)-1′,2-bis(diphenylphosphino)ferrocenyl]ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.