Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific™

Catalog No. AAL19686MC
Change view
Click to view available options
Quantity:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19686MC 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19686MC Supplier Thermo Scientific Chemicals Supplier No. L19686MC

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine is the raw material of asymmetric hydrogenation chiral ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Specifications

CAS 55650-58-3
Molecular Formula C26H28FeNP
Quantity 100mg
MDL Number MFCD00075285
Synonym s-+-n,n-dimethyl-1-2-diphenylphosphino ferrocenylethylamine, s-+-n,n-dimethyl-1-2-diphenylphosphino ferrocenyl ethylamine, s-+-n,n-dimethyl-1-r-2-diphenylphosphino ferrocenyl ethylamine, s-ppfa, +-s-n,n-dimethyl-1-r-2-diphenylphosphino ferrocenyl ethylamine
Optical Rotation +355° (c=0.6 in Ethanol)
Formula Weight 441.34
Percent Purity 97%
Chemical Name or Material (S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.