Learn More
Apexbio Technology LLC S-NAPROXEN-25G
SDP
Catalog No. 502240537
Encompass_Preferred
(S)-Naproxen (CAS No 22204-53-1) is a nonsteroidal anti-inflammatory drug (NSAID) primarily studied in the field of inflammation and pain management It originates from synthetic chemical processes and acts by inhibiting the cyclooxygenase (COX) enzymes specifically targeting COX-1 and COX-2 pathways which are crucial in the biosynthesis of prostaglandins This inhibition results in reduced inflammation and pain signaling In vitro (S)-Naproxen demonstrates activity in the nanomolar to low micromolar range It is commonly employed in various research applications including cell models to investigate inflammatory pathways animal studies to evaluate efficacy and pharmacokinetics and drug screening to explore potential therapeutic effects Experimental concentrations typically depend on the specific research design
This item is not returnable. View return policy
Catalog No. 50-224-0537 Supplier Apexbio Technology LLC Supplier No. C575525000
(S)-Naproxen (CAS No 22204-53-1) is a nonsteroidal anti-inflammatory drug (NSAID) primarily studied in the field of inflammation and pain management It originates from synthetic chemical processes and acts by inhibiting the cyclooxygenase (COX) enzymes specifically targeting COX-1 and COX-2 pathways which are crucial in the biosynthesis of prostaglandins This inhibition results in reduced inflammation and pain signaling In vitro (S)-Naproxen demonstrates activity in the nanomolar to low micromolar range It is commonly employed in various research applications including cell models to investigate inflammatory pathways animal studies to evaluate efficacy and pharmacokinetics and drug screening to explore potential therapeutic effects Experimental concentrations typically depend on the specific research design