Learn More
(S)-Naproxen methyl ester Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 26159-35-3 |
| Linear Formula | C15H16O3 |
| MDL Number | MFCD12031841 |
| Synonym | Naproxen Impurity E (PhEur); (2 S)-2-(6-Methoxy-2-naphthyl)propionic acid methyl ester; Methyl (alphaS)-6-methoxy-alpha-methyl-2-naphthaleneacetate; Methyl (2 S)-2-(6-methoxynaphthalen-2-yl)propanoate |
| Type | Pharmaceutical Impurity Standard |
| Quantity | 50 mg |
| Formula Weight | 244.29 |
| Percent Purity | ≥98.0% (HPLC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.