Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole], TCI America™
SDP

Catalog No. A5567100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5567100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5567100MG Supplier TCI America Supplier No. A5567100MG
Add to Cart
Add to Cart

[HPLC Labeling Reagent for e.e.Determination]

Chemical Identifiers

CAS 159717-68-7
Molecular Formula C11H9ClN4O4
Molecular Weight (g/mol) 296.67
MDL Number MFCD00191512
InChI Key XBQYXPYABMCADZ-UHFFFAOYNA-N
Synonym (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 44630076
IUPAC Name 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidine-2-carbonyl chloride
SMILES [O-][N+](=O)C1=CC=C(N2CCCC2C(Cl)=O)C2=NON=C12

Specifications

Quantity 100 mg
Formula Weight 296.67
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.