Learn More
TARGETMOL CHEMICALS INC S-Niguldipine hydrochloride
SDP
Catalog No. 502876054
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (S)-(+)-Niguldipine hydrochloride is a selective alpha1A-AR antagonist used in the study of depression and epilepsy. Purity 98.06%
Catalog No. 50-287-6054 Supplier TARGETMOL CHEMICALS INC Supplier No. T233015MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (S)-(+)-Niguldipine hydrochloride is a selective alpha1A-AR antagonist used in the study of depression and epilepsy. Purity 98.06%