Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Nimodipine | 77940-93-3 | 99.9% | 418.44 | 1 MG
SDP

Catalog No. 5000365236
Encompass_Preferred

(S)-Nimodipine is an enantiomer of Nimodipine, a light-sensitive dihydropyridine calcium antagonist. This orally active and well-tolerated compound is utilized in the research of cerebrovascular disorders.

  • Enantiomer of Nimodipine
  • Orally active
  • Dihydropyridine calcium antagonist
  • Target: Calcium channel
  • Pathway: Membrane transporter/ion channel; neuronal signaling
  • Appearance: White to off-white solid
  • Purity: 99.9%
  • Molecular formula: C21H26N2O7

Catalog No. 50-003-65236 Supplier Medchemexpress LLC Supplier No. 1MGHYB0265B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Nimodipine is an enantiomer of Nimodipine, a light-sensitive dihydropyridine calcium antagonist. This orally active and well-tolerated compound is utilized in the research of cerebrovascular disorders.

  • Enantiomer of Nimodipine
  • Orally active
  • Dihydropyridine calcium antagonist
  • Target: Calcium channel
  • Pathway: Membrane transporter/ion channel; neuronal signaling
  • Appearance: White to off-white solid
  • Purity: 99.9%
  • Molecular formula: C21H26N2O7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.