Learn More
Medchemexpress LLC (S)-Nimodipine | 77940-93-3 | 99.9% | 418.44 | 25 MG
SDP
Catalog No. 5000366033
Encompass_Preferred
(S)-Nimodipine is an enantiomer of Nimodipine. This orally active dihydropyridine calcium antagonist is well-tolerated and light-sensitive, making it suitable for research into cerebrovascular disorders.
  • Enantiomer of Nimodipine
  • Orally active
  • Well-tolerated
  • Light-sensitive dihydropyridine calcium antagonist
  • Used for research of cerebrovascular disorders
  • Targets Calcium Channel
  • Involved in Membrane Transporter/Ion Channel pathway
  • Involved in Neuronal Signaling pathway
Catalog No. 50-003-66033 Supplier Medchemexpress LLC Supplier No. 25MGHYB0265B
(S)-Nimodipine is an enantiomer of Nimodipine. This orally active dihydropyridine calcium antagonist is well-tolerated and light-sensitive, making it suitable for research into cerebrovascular disorders.
  • Enantiomer of Nimodipine
  • Orally active
  • Well-tolerated
  • Light-sensitive dihydropyridine calcium antagonist
  • Used for research of cerebrovascular disorders
  • Targets Calcium Channel
  • Involved in Membrane Transporter/Ion Channel pathway
  • Involved in Neuronal Signaling pathway