Learn More
Cayman Chemical (S)-nitro-Blebbistatin, Purity: ≥98%, CAS: 856925-75-2, 10 mg
SDP
Catalog No. 502954335
Encompass_Preferred
A more stable form of (–)-blebbistatin, the active form of blebbistatin; has the same stereochemistry as the active (–)-blebbistatin enantiomer
Catalog No. 50-295-4335 Supplier Cayman Chemical Supplier No. 1389110 MG
A more stable form of (–)-blebbistatin, the active form of blebbistatin; has the same stereochemistry as the active (–)-blebbistatin enantiomer