Learn More
Cayman Chemical (S)-nitro-Blebbistatin, 856925-75-2, 1 mg
SDP
Supplier:  Cayman Chemical 138911
Encompass_Preferred
Molecular Formula C17H13N3O4, Purity ≥98%, Formula Weight 323.3, Synonyms: (3aS)-1,2,3,3a-tetrahydro-3a-hydroxy-7-nitro-1-phenyl-4H-pyrrolo[2,3-b]quinolin-4-one, S-(−)-7-Desmethyl-8-nitro Blebbistatin; A more stable form of ()-blebbistatin, the active form of blebbistatin; has the same stereochemistry as the active ()-blebbistatin enantiomer.
Catalog No. NC0664123