Learn More
Medchemexpress LLC HY-121526 25mg , S-Nitroso-N-acetyl-DL-penicillamine CAS:67776-06-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12152625MG
Encompass_Preferred
HY-121526 25mg S-Nitroso-N-acetyl-DL-penicillamine CAS: 67776-06-1 S-Nitroso-N-acetyl-DL-penicillamine (SNAP) is a nitric oxide donor and acts as a stable inhibitor of platelet aggregation[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0609