Learn More
Apexbio Technology LLC S-Nitroso-N-acetyl-DL-penicillamine 67776-06-1 50mg
SDP
Catalog No. 5000096287
Encompass_Preferred
S-Nitroso-N-acetyl-DL-penicillamine (SNAP CAS 67776-06-1) is a low-molecular-weight nitric oxide (NO) donor that facilitates the sustained release of NO in biological systems By elevating NO levels SNAP stimulates soluble guanylate cyclase activity resulting in increased cyclic guanosine monophosphate (cGMP) production and subsequent modulation of downstream signaling pathways These actions contribute to vascular smooth muscle relaxation and vasodilation SNAP is widely employed in experimental studies to investigate NO-mediated mechanisms including inhibition of platelet aggregation and regulation of inflammatory responses and serves as a valuable reagent for probing NO signaling in cultured cells
Catalog No. 50-000-96287 Supplier Apexbio Technology LLC Supplier No. A462150
S-Nitroso-N-acetyl-DL-penicillamine (SNAP CAS 67776-06-1) is a low-molecular-weight nitric oxide (NO) donor that facilitates the sustained release of NO in biological systems By elevating NO levels SNAP stimulates soluble guanylate cyclase activity resulting in increased cyclic guanosine monophosphate (cGMP) production and subsequent modulation of downstream signaling pathways These actions contribute to vascular smooth muscle relaxation and vasodilation SNAP is widely employed in experimental studies to investigate NO-mediated mechanisms including inhibition of platelet aggregation and regulation of inflammatory responses and serves as a valuable reagent for probing NO signaling in cultured cells