Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S-nitroso-N-acetyl-DL-penicillamine | 67776-06-1 | 98.8% | 100 MG
SDP

Catalog No. 5000232266
Encompass_Preferred

S-nitroso-N-acetyl-DL-penicillamine (SNAP) is a nitric oxide donor used in biochemical and physiological research to release NO. It acts as a stable inhibitor of platelet aggregation and is supplied as a powdered reagent for laboratory studies.

  • Acts as a nitric oxide donor and inhibitor of platelet aggregation.
  • Molecular formula C7H12N2O4S and molecular weight 220.25 g/mol.
  • Purity 98.8% (as supplied).
  • Powder form, commonly available in small research pack sizes such as 100 MG.
  • Stored at low temperature to maintain stability (powder storage at -20°C recommended).
  • Used in studies of nitric oxide signaling, platelet function, and related assays.

Catalog No. 50-002-32266 Supplier Medchemexpress LLC Supplier No. HY121526100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-nitroso-N-acetyl-DL-penicillamine (SNAP) is a nitric oxide donor used in biochemical and physiological research to release NO. It acts as a stable inhibitor of platelet aggregation and is supplied as a powdered reagent for laboratory studies.

  • Acts as a nitric oxide donor and inhibitor of platelet aggregation.
  • Molecular formula C7H12N2O4S and molecular weight 220.25 g/mol.
  • Purity 98.8% (as supplied).
  • Powder form, commonly available in small research pack sizes such as 100 MG.
  • Stored at low temperature to maintain stability (powder storage at -20°C recommended).
  • Used in studies of nitric oxide signaling, platelet function, and related assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.