Learn More
Medchemexpress LLC S-nitroso-N-acetyl-DL-penicillamine | 67776-06-1 | 98.8% | 5 MG
SDP
Catalog No. 5000231234
Encompass_Preferred
S-nitroso-N-acetyl-DL-penicillamine (SNAP) is a nitric oxide donor commonly used in biochemical research to provide controlled NO release for studies of vascular function, signaling pathways, and platelet aggregation. It is supplied as a high-purity research chemical for in vitro experiments.
  • Releases nitric oxide under physiological conditions.
  • Suitable for studies of vasodilation and platelet aggregation.
  • High purity (98.8% by HPLC) as confirmed by COA.
  • Defined molecular properties: 220.25 g/mol, formula C7H12N2O4S.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-31234 Supplier Medchemexpress LLC Supplier No. HY1215265MG
S-nitroso-N-acetyl-DL-penicillamine (SNAP) is a nitric oxide donor commonly used in biochemical research to provide controlled NO release for studies of vascular function, signaling pathways, and platelet aggregation. It is supplied as a high-purity research chemical for in vitro experiments.
  • Releases nitric oxide under physiological conditions.
  • Suitable for studies of vasodilation and platelet aggregation.
  • High purity (98.8% by HPLC) as confirmed by COA.
  • Defined molecular properties: 220.25 g/mol, formula C7H12N2O4S.
  • Available in small research pack sizes for laboratory use.