Learn More
Medchemexpress LLC S-nitroso-N-acetylpenicillamine | 79032-48-7 | MFCD00153852 | 95.0% | 220.25 | C7H12N2O4S | 10 MG
SDP
Catalog No. 5000211437
Encompass_Preferred
D-SNAP (S-nitroso-N-acetylpenicillamine) is an S-nitrosothiol nitric oxide donor used in biochemical and cell biology research to deliver controlled NO release and to probe NO-mediated signaling and oxidative stress. It is provided as a solid for preparation of stock solutions and requires light-protected, inert storage for stability.
  • Generates nitric oxide under physiological conditions.
  • Typical purity 95.0%.
  • Available in small milligram quantities for research use.
  • Stable when stored at -20°C, protected from light and under nitrogen.
  • Provides controlled and reproducible NO release for cellular assays.
Catalog No. 50-002-11437 Supplier Medchemexpress LLC Supplier No. HY10101310MG
D-SNAP (S-nitroso-N-acetylpenicillamine) is an S-nitrosothiol nitric oxide donor used in biochemical and cell biology research to deliver controlled NO release and to probe NO-mediated signaling and oxidative stress. It is provided as a solid for preparation of stock solutions and requires light-protected, inert storage for stability.
  • Generates nitric oxide under physiological conditions.
  • Typical purity 95.0%.
  • Available in small milligram quantities for research use.
  • Stable when stored at -20°C, protected from light and under nitrogen.
  • Provides controlled and reproducible NO release for cellular assays.