Learn More
Medchemexpress LLC S-nitroso-N-acetylpenicillamine | 79032-48-7 | MFCD00272624 | 95.0% | 220.25 | C7H12N2O4S | 5 MG
SDP
Catalog No. 5000211899
Encompass_Preferred
D-SNAP (S-nitroso-N-acetylpenicillamine) is a nitrosothiol reagent used as a nitric oxide donor and fluorescent probe in biochemical and cellular research. Provided in a milligram-scale vial at research-grade purity, it is intended for laboratory assays and should be handled and stored per the safety data sheet.
  • Acts as a nitric oxide donor for in vitro and cellular experiments.
  • Functions as a fluorescent reagent for detection and labeling applications.
  • High purity suitable for research use.
  • Molecular weight 220.25 g/mol and formula C7H12N2O4S.
  • Available in milligram-scale vials for small-scale experiments.
Catalog No. 50-002-11899 Supplier Medchemexpress LLC Supplier No. HY1010135MG
D-SNAP (S-nitroso-N-acetylpenicillamine) is a nitrosothiol reagent used as a nitric oxide donor and fluorescent probe in biochemical and cellular research. Provided in a milligram-scale vial at research-grade purity, it is intended for laboratory assays and should be handled and stored per the safety data sheet.
  • Acts as a nitric oxide donor for in vitro and cellular experiments.
  • Functions as a fluorescent reagent for detection and labeling applications.
  • High purity suitable for research use.
  • Molecular weight 220.25 g/mol and formula C7H12N2O4S.
  • Available in milligram-scale vials for small-scale experiments.