Learn More
AdipoGen S-Nitrosoglutathione
SDP
Supplier:  AdipoGen CDXN0262M050
Encompass_Preferred
Chemical. CAS 57564-91-7. Formula C10H16N4O7S. Molecular Weight 336.32. S-Nitroso-L-glutathione is an S-nitrosothiol NO donor based on naturally-occurring glutathione. It is a potent smooth muscle relaxant and inhibitor of platelet aggregation. Its stability in solution varies from seconds to hours depending on temperature, buffer composition and metal content.
Catalog No. 50-256-0373