Learn More
Selleck Chemical LLC S-Nitrosoglutathione E4443-25MG
SDP
Catalog No. 5000039705
Encompass_Preferred
S-nitrosoglutathione (GSNO RVC-588 S-Nitroso-L-glutathione Nitrosoglutathione) is an amino acid and nitric oxide donor that under physiological conditions spontaneously releases nitric oxide S-nitrosoglutathione (GSNO) selectively inhibits platelet and NF- B activation It induces apoptosis in T cells in low concentrations ( 0 6 mM) while protecting T cells from apoptosis in higher concentrations (1-2 mM)
Catalog No. 50-000-39705 Supplier Selleck Chemical LLC Supplier No. E444325MG
S-nitrosoglutathione (GSNO RVC-588 S-Nitroso-L-glutathione Nitrosoglutathione) is an amino acid and nitric oxide donor that under physiological conditions spontaneously releases nitric oxide S-nitrosoglutathione (GSNO) selectively inhibits platelet and NF- B activation It induces apoptosis in T cells in low concentrations ( 0 6 mM) while protecting T cells from apoptosis in higher concentrations (1-2 mM)