Learn More
Abcam S-Nitrosoglutathione (SNOG), NO donor, 50MG
Catalog No. 502707717
Encompass_Preferred
MW 336.32 Da, Purity >=98%. A carrier of nitric oxide. Breaks down at physiological pH to produce nitric oxide. Relaxes smooth muscle and inhibits platelet aggregation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7717 Supplier Abcam Supplier No. AB12028550MG
MW 336.32 Da, Purity >=98%. A carrier of nitric oxide. Breaks down at physiological pH to produce nitric oxide. Relaxes smooth muscle and inhibits platelet aggregation.
The product is subject to the following: Abcam Restricted Use Statement