Learn More
Teknova SOB Broth. 454g.
SDP
Catalog No. 50843052
Encompass
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.
Catalog No. 50-843-052 Supplier Teknova Supplier No. S0210
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.