Learn More
Teknova SOB Medium, 1000mL, Sterile.
SDP
Catalog No. 50843103
Encompass
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.
This item is not returnable. View return policy
Catalog No. 50-843-103 Supplier Teknova Supplier No. S1615
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.