Learn More
Teknova SOC Broth, 227 grams.
SDP
Catalog No. 50843063
Encompass
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.
Catalog No. 50-843-063 Supplier Teknova Supplier No. S0225
SOB (Super Optimal Broth) Medium is a nutrient rich medium for cultivation ofE. coli, often used to give high transformation efficiency. SOC Medium is used for providing nutrients and supporting the recovery of Escherichia coli competent cells following transformation.