Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-O-Desmethylraclopride 98.0+%, TCI America™
SDP

Catalog No. D4134200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4134200MG 200 mg
D41341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4134200MG Supplier TCI America Supplier No. D4134200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119670-11-0
Molecular Formula C14H18Cl2N2O3
Molecular Weight (g/mol) 333.209
MDL Number MFCD02179385
InChI Key DPQOMEPZWBXAMA-QMMMGPOBSA-N
Synonym (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dihydroxybenzamide
PubChem CID 10947574
IUPAC Name 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2,6-dihydroxybenzamide
SMILES CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2O)Cl)Cl)O

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 200 mg
Formula Weight 333.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-O-Desmethylraclopride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.