Learn More
Hampton Research S.O. PHOSPHATE #01 PH 5.0
SDP
Catalog No. 50563168
Encompass
HR2-251-01/StockOptions Phosphate #01, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.0 (0.980 M Sodium phosphate monobasic monohydrate, 0.020 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-168 Supplier Hampton Research Supplier No. HR225101
HR2-251-01/StockOptions Phosphate #01, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.0 (0.980 M Sodium phosphate monobasic monohydrate, 0.020 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.