Learn More
Hampton Research S.O. PHOSPHATE #03 PH 5.4
SDP
Catalog No. 50563170
Encompass
HR2-251-03/StockOptions Phosphate #03, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.4 (0.948 M Sodium phosphate monobasic monohydrate, 0.052 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-170 Supplier Hampton Research Supplier No. HR225103
HR2-251-03/StockOptions Phosphate #03, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.4 (0.948 M Sodium phosphate monobasic monohydrate, 0.052 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.