Learn More
Hampton Research S.O. PHOSPHATE #05 PH 5.8
SDP
Catalog No. 50563172
Encompass
HR2-251-05/StockOptions Phosphate #05, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.8 (0.840 M Sodium phosphate monobasic monohydrate, 0.160 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-172 Supplier Hampton Research Supplier No. HR225105
HR2-251-05/StockOptions Phosphate #05, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 5.8 (0.840 M Sodium phosphate monobasic monohydrate, 0.160 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.