Learn More
Hampton Research S.O. PHOSPHATE #06 PH 6.0
SDP
Catalog No. 50563173
Encompass
HR2-251-06/StockOptions Phosphate #06, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.0 (0.764 M Sodium phosphate monobasic monohydrate, 0.236 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-173 Supplier Hampton Research Supplier No. HR225106
HR2-251-06/StockOptions Phosphate #06, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.0 (0.764 M Sodium phosphate monobasic monohydrate, 0.236 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.