Learn More
Hampton Research S.O. PHOSPHATE #07 PH 6.2
SDP
Catalog No. 50563174
Encompass
HR2-251-07/StockOptions Phosphate #07, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.2 (0.684 M Sodium phosphate monobasic monohydrate, 0.316 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-174 Supplier Hampton Research Supplier No. HR225107
HR2-251-07/StockOptions Phosphate #07, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.2 (0.684 M Sodium phosphate monobasic monohydrate, 0.316 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.