Learn More
Hampton Research S.O. PHOSPHATE #08 PH 6.4
SDP
Catalog No. 50563175
Encompass
HR2-251-08/StockOptions Phosphate #08, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.4 (0.592 M Sodium phosphate monobasic monohydrate, 0.408 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-175 Supplier Hampton Research Supplier No. HR225108
HR2-251-08/StockOptions Phosphate #08, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.4 (0.592 M Sodium phosphate monobasic monohydrate, 0.408 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.