Learn More
Hampton Research S.O. PHOSPHATE #10 PH 6.8
SDP
Catalog No. 50563177
Encompass
HR2-251-10/StockOptions Phosphate #10, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.8 (0.412 M Sodium phosphate monobasic monohydrate, 0.588 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-177 Supplier Hampton Research Supplier No. HR225110
HR2-251-10/StockOptions Phosphate #10, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.8 (0.412 M Sodium phosphate monobasic monohydrate, 0.588 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.