Learn More
Hampton Research S.O. PHOSPHATE #11 PH 7.0
SDP
Catalog No. 50563178
Encompass
HR2-251-11/StockOptions Phosphate #11, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.0 (0.324 M Sodium phosphate monobasic monohydrate, 0.676 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-178 Supplier Hampton Research Supplier No. HR225111
HR2-251-11/StockOptions Phosphate #11, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.0 (0.324 M Sodium phosphate monobasic monohydrate, 0.676 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.