Learn More
Hampton Research S.O. PHOSPHATE #12 PH 7.2
SDP
Catalog No. 50563179
Encompass
HR2-251-12/StockOptions Phosphate #12, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.2 (0.240 M Sodium phosphate monobasic monohydrate, 0.760 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-179 Supplier Hampton Research Supplier No. HR225112
HR2-251-12/StockOptions Phosphate #12, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.2 (0.240 M Sodium phosphate monobasic monohydrate, 0.760 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.