Learn More
Hampton Research S.O. PHOSPHATE #13 PH 7.4
SDP
Catalog No. 50563180
Encompass
HR2-251-13/StockOptions Phosphate #13, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.4 (0.168 M Sodium phosphate monobasic monohydrate, 0.832 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-180 Supplier Hampton Research Supplier No. HR225113
HR2-251-13/StockOptions Phosphate #13, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.4 (0.168 M Sodium phosphate monobasic monohydrate, 0.832 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.