Learn More
Hampton Research S.O. PHOSPHATE #14 PH 7.6
SDP
Catalog No. 50563181
Encompass
HR2-251-14/StockOptions Phosphate #14, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.6 (0.108 M Sodium phosphate monobasic monohydrate, 0.892 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-181 Supplier Hampton Research Supplier No. HR225114
HR2-251-14/StockOptions Phosphate #14, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.6 (0.108 M Sodium phosphate monobasic monohydrate, 0.892 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.