Learn More
Hampton Research S.O. PHOSPHATE #15 PH 7.8
SDP
Catalog No. 50563182
Encompass
HR2-251-15/StockOptions Phosphate #15, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.8 (0.068 M Sodium phosphate monobasic monohydrate, 0.932 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-182 Supplier Hampton Research Supplier No. HR225115
HR2-251-15/StockOptions Phosphate #15, 1.0 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 7.8 (0.068 M Sodium phosphate monobasic monohydrate, 0.932 M Potassium phosphate dibasic), 185 ml. Please note items from this supplier are non-returnable.