Learn More
Hampton Research S.O. SUCCINIC ACID #01 PH 4.3
SDP
Catalog No. 50563123
Encompass
HR2-249-01/StockOptions Succinic Acid #01, 1.0 M Succinic acid pH 4.3, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-123 Supplier Hampton Research Supplier No. HR224901
HR2-249-01/StockOptions Succinic Acid #01, 1.0 M Succinic acid pH 4.3, 185 ml. Please note items from this supplier are non-returnable.