Learn More
Hampton Research S.O. SUCCINIC ACID #06 PH 4.8
SDP
Catalog No. 50563128
Encompass
HR2-249-06/StockOptions Succinic Acid #06, 1.0 M Succinic acid pH 4.8, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-128 Supplier Hampton Research Supplier No. HR224906
HR2-249-06/StockOptions Succinic Acid #06, 1.0 M Succinic acid pH 4.8, 185 ml. Please note items from this supplier are non-returnable.