Learn More
Hampton Research S.O. SUCCINIC ACID #07 PH 4.9
SDP
Catalog No. 50563129
Encompass
HR2-249-07/StockOptions Succinic Acid #07, 1.0 M Succinic acid pH 4.9, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-129 Supplier Hampton Research Supplier No. HR224907
HR2-249-07/StockOptions Succinic Acid #07, 1.0 M Succinic acid pH 4.9, 185 ml. Please note items from this supplier are non-returnable.