Learn More
Hampton Research S.O. SUCCINIC ACID #08 PH 5.0
SDP
Catalog No. 50563130
Encompass
HR2-249-08/StockOptions Succinic Acid #08, 1.0 M Succinic acid pH 5.0, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-130 Supplier Hampton Research Supplier No. HR224908
HR2-249-08/StockOptions Succinic Acid #08, 1.0 M Succinic acid pH 5.0, 185 ml. Please note items from this supplier are non-returnable.