Learn More
Hampton Research S.O. SUCCINIC ACID #14 PH 5.6
SDP
Catalog No. 50563136
Encompass
HR2-249-14/StockOptions Succinic Acid #14, 1.0 M Succinic acid pH 5.6, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-136 Supplier Hampton Research Supplier No. HR224914
HR2-249-14/StockOptions Succinic Acid #14, 1.0 M Succinic acid pH 5.6, 185 ml. Please note items from this supplier are non-returnable.