Learn More
Hampton Research S.O. SUCCINIC ACID #15 PH 5.7
SDP
Catalog No. 50563137
Encompass
HR2-249-15/StockOptions Succinic Acid #15, 1.0 M Succinic acid pH 5.7, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-137 Supplier Hampton Research Supplier No. HR224915
HR2-249-15/StockOptions Succinic Acid #15, 1.0 M Succinic acid pH 5.7, 185 ml. Please note items from this supplier are non-returnable.