Learn More
Hampton Research S.O. SUCCINIC ACID #18 PH 6.0
SDP
Catalog No. 50563140
Encompass
HR2-249-18/StockOptions Succinic Acid #18, 1.0 M Succinic acid pH 6.0, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-140 Supplier Hampton Research Supplier No. HR224918
HR2-249-18/StockOptions Succinic Acid #18, 1.0 M Succinic acid pH 6.0, 185 ml. Please note items from this supplier are non-returnable.