Learn More
Hampton Research S.O. SUCCINIC ACID #19 PH 6.1
SDP
Catalog No. 50563141
Encompass
HR2-249-19/StockOptions Succinic Acid #19, 1.0 M Succinic acid pH 6.1, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-141 Supplier Hampton Research Supplier No. HR224919
HR2-249-19/StockOptions Succinic Acid #19, 1.0 M Succinic acid pH 6.1, 185 ml. Please note items from this supplier are non-returnable.