Learn More
Hampton Research S.O. SUCCINIC ACID #21 PH 6.3
SDP
Catalog No. 50563143
Encompass
HR2-249-21/StockOptions Succinic Acid #21, 1.0 M Succinic acid pH 6.3, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-143 Supplier Hampton Research Supplier No. HR224921
HR2-249-21/StockOptions Succinic Acid #21, 1.0 M Succinic acid pH 6.3, 185 ml. Please note items from this supplier are non-returnable.