Learn More
Hampton Research S.O. SUCCINIC ACID #23 PH 6.5
SDP
Catalog No. 50563145
Encompass
HR2-249-23/StockOptions Succinic Acid #23, 1.0 M Succinic acid pH 6.5, 185 ml. Please note items from this supplier are non-returnable.
Catalog No. 50-563-145 Supplier Hampton Research Supplier No. HR224923
HR2-249-23/StockOptions Succinic Acid #23, 1.0 M Succinic acid pH 6.5, 185 ml. Please note items from this supplier are non-returnable.